Anti-B2M Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-141-0100
RL200401141010EA 551.64 USD
RL200401141010
Anti-B2M Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: B2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR