Anti-TERF2 Mouse Monoclonal Antibody [clone: 4A794.15]

Supplier: Rockland Immunochemical
200-301-I34
RL200301I34EA 702.09 USD
RL200301I34
Anti-TERF2 Mouse Monoclonal Antibody [clone: 4A794.15]
Antibodies
Anti-TRF2 Antibody is suitable for use in ChIP, ELISA, Flow (Intracellular) Cytometry, IF/ICC, Immunochemistry(frozen), IP, and WB. Expect a band approximately 55.6 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 2 µg/ml; ChIP: User Optimized; Immunofluorescence: User Optimized; Western Blot: 2-4 µg/ml; Immunochemistry: 5 µg/ml; Immunoprecipitation: 2 µg/10^6 cells

Type: Primary
Antigen: TERF2
Clonality: Monoclonal
Clone: 4A794.15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR