Anti-TLR9 Mouse Monoclonal Antibody [clone: 26C593.2]

Supplier: Rockland Immunochemical
200-301-I31
RL200301I31EA 687.46 USD
RL200301I31
Anti-TLR9 Mouse Monoclonal Antibody [clone: 26C593.2]
Antibodies
Anti-TLR9 Antibody is suitable for use inCM, Flow (CS), Flow (I), IF/ICC, Immunochemistry(frozen), Immunochemistry(p) and WB. Expect a band approximately 113kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1-5 µg/ml; Immunochemistry: 10-20 µg/ml (frozen)

Type: Primary
Antigen: TLR9
Clonality: Monoclonal
Clone: 26C593.2
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR