Anti-TLR8 Mouse Monoclonal Antibody [clone: 44C143]

Supplier: Rockland Immunochemical
200-301-I30
RL200301I30EA 643.58 USD
RL200301I30
Anti-TLR8 Mouse Monoclonal Antibody [clone: 44C143]
Antibodies
Anti-TLR8 Antibody is suitable for use in Flow (CS), Flow (I), Immunochemistry(p) and WB.Expect a band approximately 121kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1-3 µg/ml; Immunochemistry: 5 µg/ml

Type: Primary
Antigen: TLR8
Clonality: Monoclonal
Clone: 44C143
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR