Anti-TLR4 Mouse Monoclonal Antibody [clone: 76B357.1]

Supplier: Rockland Immunochemical
200-301-I25
RL200301I25EA 687.46 USD
RL200301I25
Anti-TLR4 Mouse Monoclonal Antibody [clone: 76B357.1]
Antibodies
Anti-TLR4 Antibody is suitable for use in Flow (CS), Flow (I), IF, Immunochemistry(p) and WB. Expect a band approximately 95kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 1:10 - 1:2000; Flow Cytometry: 0.5 - 2.0 µg/10^6 cells; Western Blot: 1 - 3 µg/ml; Immunochemistry: 5 µg/ml

Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: 76B357.1
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR