Anti-PARP1 Mouse Monoclonal Antibody [clone: 194C1439]

Supplier: Rockland Immunochemical
200-301-I03
RL200301I03EA 575.67 USD
RL200301I03
Anti-PARP1 Mouse Monoclonal Antibody [clone: 194C1439]
Antibodies
Anti-PARP antibodyis suitable for use in Flow (I) and WB. Expect a band approximately 89kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Flow Cytometry: 1-3 µg/test; Western Blot: 1-2 µg/ml

Type: Primary
Antigen: PARP1
Clonality: Monoclonal
Clone: 194C1439
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR