Anti-Dnmt3a Mouse Monoclonal Antibody [clone: 64B1446]

Supplier: Rockland Immunochemical
200-301-H70
RL200301H70EA 702.09 USD
RL200301H70
Anti-Dnmt3a Mouse Monoclonal Antibody [clone: 64B1446]
Antibodies
Anti-Dnmt3a Antibody is suitable for use in ChIP, IF/ICC, Immunochemistry(p) and WB. Expect a band approximately 120 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 5 µg/ml; Western Blot: 2 µg/ml

Type: Primary
Antigen: Dnmt3a
Clonality: Monoclonal
Clone: 64B1446
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR