Anti-CASP8 Mouse Monoclonal Antibody [clone: 90A992]

Supplier: Rockland Immunochemical
200-301-H64
RL200301H64EA 614.33 USD
RL200301H64
Anti-CASP8 Mouse Monoclonal Antibody [clone: 90A992]
Antibodies
Anti-Caspase-8 antibody is suitable for use by Flow (I), Immunochemistry(p), and WB. Expect a band approximately 55 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Flow Cytometry: 0.1 - 0.5 µg/ml; Western Blot: 0.5 - 2 µg/ml; Immunochemistry: 5 µg/ml

Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: 90A992
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR