Anti-BIN1 Mouse Monoclonal Antibody [clone: 2F11]

Supplier: Rockland Immunochemical
200-301-E63
RL200301E63EA 565.22 USD
RL200301E63
Anti-BIN1 Mouse Monoclonal Antibody [clone: 2F11]
Antibodies
Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR