Anti-BIN1 Mouse Monoclonal Antibody [clone: 99F]

Supplier: Rockland Immunochemical
200-301-E61S
RL200301E61SEA 151.49 USD
RL200301E61S
Anti-BIN1 Mouse Monoclonal Antibody [clone: 99F]
Antibodies
Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 99F
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR