Anti-CAR1 Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-1654-0100
RL20016540100EA 531.79 USD
RL20016540100
Anti-CAR1 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CAR1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR