Anti-PKM Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-1178-0100 200-1178S
RL20011780100EA 551.64 USD
RL20011780100 RL2001178S
Anti-PKM Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:500 - 1:2000; Other Dilution: User Optimized

Type: Primary
Antigen: PKM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR