Anti-PGA Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-1176-0100 200-1176S
RL20011760100EA 551.64 USD
RL20011760100 RL2001176S
Anti-PGA Goat Polyclonal Antibody
Antibodies
Anti-Pepsin antibody is suitable for western blotting to detect a single band of the expected apparent molecular weight and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:50:000 - 1:200000; Western Blot: 1:5000 - 1:20000; Immunochemistry: 1:2000 - 1:10000; Immunoprecipitation: 1:100; Other Dilution: User Optimized

Type: Primary
Antigen: PGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR