Anti-GLNA Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-101-238S
RL200101238SEA 151.49 USD
RL200101238S
Anti-GLNA Goat Polyclonal Antibody
Antibodies
Anti-Glutamine Synthetase antibody has been assayed against 1.0 µg of Glutamine Synthetase [Microbial] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG [H&L] (Rabbit) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100; Other Dilution: User Optimized

Type: Primary
Antigen: glnA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Brevibacterium
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR