Anti-PGMB Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-101-237S
RL200101237SEA 151.49 USD
RL200101237S
Anti-PGMB Goat Polyclonal Antibody
Antibodies
BETA-PHOSPHOGLUCOMUTASE Antibody has been assayed against 1.0 µg of b-Phosphoglucomutase [Lactococcus lacti] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG [H&L] (Rabbit) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000; Other Dilution: User Optimized

Type: Primary
Antigen: PGMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Lactococcus lacti
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR