Anti-amyA Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-101-234S
RL200101234SEA 151.49 USD
RL200101234S
Anti-amyA Goat Polyclonal Antibody
Antibodies
GLUCOAMYLASE Antibody has been assayed against 1.0 µg of Glucoamylase [Rhizopus] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG [H&L] (Goat) and (ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000; Other Dilution: User Optimized

Type: Primary
Antigen: amyA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rhizopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR