Anti-Sucrose Oxidase Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-101-231-0100
RL200101231010EA 551.64 USD
RL200101231010
Anti-Sucrose Oxidase Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Sucrose Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: E. Coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR