Anti-FOLR1 Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-101-148S
RL200101148SEA 151.49 USD
RL200101148S
Anti-FOLR1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FOLR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR