Anti-BAX Rabbit Monoclonal Antibody [clone: SP47]

Supplier: Rockland Immunochemical
200-C01-B34
RL200-C01-B34EA 520.3 USD
RL200-C01-B34
Anti-BAX Rabbit Monoclonal Antibody [clone: SP47]
Antibodies
This monoclonal antibody is suitable for ELISA, immunochemistry and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 21 kDa in size corresponding to BAX Protein by western blotting.

Recommended Dilutions: ELISA: 1:20000; Flow Cytometry: 1:100; Western Blot: 1:500 – 1:2000; Immunochemistry: 1:50;
Do not freeze!

Type: Primary
Antigen: BAX
Clonality: Monoclonal
Clone: SP47
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR