Anti-NEFL Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical
200-901-D83
RL200-901-D83EA 578.81 USD
RL200-901-D83
Anti-NEFL Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NEFL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Mouse , Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR