Anti-CD8A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 53-6.7]

Supplier: Rockland Immunochemical
200-526-N77
RL200-526-N77EA 436.72 USD
RL200-526-N77
Anti-CD8A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 53-6.7]
Antibodies
Anti-CD8a is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR