Anti-AKT2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 16G11.E8]

Supplier: Rockland Immunochemical
200-526-E71
RL200-526-E71EA 575.67 USD
RL200-526-E71
Anti-AKT2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 16G11.E8]
Antibodies
Anti-AKT2 APC Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT2 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: Flow Cytometry: 1.0 µg/ml;
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 16G11.E8
Conjugation: APC (Allophycocyanin)
Epitope: Center
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR