Anti-CD45.1 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: A20]

Supplier: Rockland Immunochemical
200-508-N85
RL200-508-N85EA 414.78 USD
RL200-508-N85
Anti-CD45.1 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: A20]
Antibodies
Anti-CD45.1 is useful for Flow Cytometry, Immunoprecipitation, and immunochemistry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR