Anti-CDKN2A Rat Polyclonal Antibody [clone: 12.A1.1]

Supplier: Rockland Immunochemical
200-501-892
RL200-501-892EA 565.22 USD
RL200-501-892
Anti-CDKN2A Rat Polyclonal Antibody [clone: 12.A1.1]
Antibodies
This antibody has been tested for use in ELISA, western blotting and immunoprecipitation. Specific conditions for western blotting reactivity should be optimized by the end user.  Expect a band approximately 19 kDa in size corresponding to mouse p19Arf.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDKN2A
Clonality: Polyclonal
Clone: 12.A1.1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR