Anti-ALP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
200-4335S
RL200-4335SEA 151.49 USD
RL200-4335S
Anti-ALP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Alkaline phosphatase has been assayed against 1.0 µg of Alkaline Phosphatase (E.coli) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:200 - 1:1000;
Do not add Sodium Azide!

Type: Primary
Antigen: ALP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR