Anti-BB-H32 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-C12S 200-401-C12
RL200-401-C12SEA 151.49 USD
RL200-401-C12S RL200-401-C12
Anti-BB-H32 Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody has been tested for use in Western blotting and ELISA. Specific conditions for reactivity should be optimized by the user. Expect a band approximately 27.1 kDa in size corresponding to Borrelia burgdorferi p35 Protein by Western blotting.

Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: BB-H32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR