Anti-dbpA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-B98S 200-401-B98
RL200-401-B98SEA 151.49 USD
RL200-401-B98S RL200-401-B98
Anti-dbpA Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody has been tested for use in Western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 18 kDa in size corresponding to Borrelia burgdorferi DbpA Protein by Western blotting.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: dbpA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR