Anti-CCL3L1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-B61 200-401-B61S
RL200-401-B61EA 551.64 USD
RL200-401-B61 RL200-401-B61S
Anti-CCL3L1 Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 7.8 kDa in size corresponding to swine CCL3L1 Protein by western blotting.

Recommended Dilutions: Western Blot: 4 µg/ml;

Type: Primary
Antigen: CCL3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR