Anti-TIRAP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-A67
RL200-401-A67EA 635.22 USD
RL200-401-A67
Anti-TIRAP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TIRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR