Anti-RFFL Rabbit Polyclonal Antibody [clone: BP53-12]

Supplier: Rockland Immunochemical
200-401-932
RL200-401-932EA 609.1 USD
RL200-401-932
Anti-RFFL Rabbit Polyclonal Antibody [clone: BP53-12]
Antibodies
This Protein A purified antibody has been tested for use in ELISA, immunochemistry and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 41 kDa in size corresponding to RFFL Protein.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 500 µl

Type: Primary
Antigen: RFFL
Clonality: Polyclonal
Clone: BP53-12
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR