Anti-S100 A1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-874
RL200-401-874EA 609.1 USD
RL200-401-874
Anti-S100 A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: S100 A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR