Anti-LYZ Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-072 200-401-072S
RL200-401-072EA 551.64 USD
RL200-401-072 RL200-401-072S
Anti-LYZ Rabbit Polyclonal Antibody
Antibodies
This purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user.  Expect a band at ~14 kDa in size corresponding to lysozyme by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: LYZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR