Anti-GFP Mouse Monoclonal Antibody (DyLight® 405) [clone: 9F9.F9]

Supplier: Rockland Immunochemical
200-346-215
RL200-346-215EA 531.79 USD
RL200-346-215
Anti-GFP Mouse Monoclonal Antibody (DyLight® 405) [clone: 9F9.F9]
Antibodies
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: ELISA: 1:20000-1:40000; FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: 1:10000-1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: GFP
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: DyLight® 405
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR