Anti-6xHis Mouse Monoclonal Antibody (DyLight® 649) [clone: 33D1.D2]

Supplier: Rockland Immunochemical
200-343-382
RL200-343-382EA 531.79 USD
RL200-343-382
Anti-6xHis Mouse Monoclonal Antibody (DyLight® 649) [clone: 33D1.D2]
Antibodies
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 33D1.D2
Conjugation: DyLight® 649
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR