Anti-DYKDDDDK Mouse Monoclonal Antibody (DyLight® 488) [clone: 29E4.G7]

Supplier: Rockland Immunochemical
200-341-383
RL200-341-383EA 498.36 USD
RL200-341-383
Anti-DYKDDDDK Mouse Monoclonal Antibody (DyLight® 488) [clone: 29E4.G7]
Antibodies
Type: Primary
Antigen: DYKDDDDK
Clonality: Monoclonal
Clone: 29E4.G7
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR