Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 17F6.B11]

Supplier: Rockland Immunochemical
200-341-268
RL200-341-268EA 543.28 USD
RL200-341-268
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 488) [clone: 17F6.B11]
Antibodies
This monoclonal antibody is suitable for immunofluorescence microscopy, FLISA and western blotting. Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:4000-1:20000; Western Blot: 1:10000 - 1:25000; Immunochemistry: 20 µg/ml;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 17F6.B11
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR