Anti-CD9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]

Supplier: Rockland Immunochemical
200-326-N70
RL200-326-N70EA 674.93 USD
RL200-326-N70
Anti-CD9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]
Antibodies
Anti-CD45RA is useful for Flow Cytometry, immunochemistry, and Western Blot. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: HI100
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR