Anti-CD8A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RPA-T8]

Supplier: Rockland Immunochemical
200-326-M64
RL200-326-M64EA 329.1 USD
RL200-326-M64
Anti-CD8A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RPA-T8]
Antibodies
Anti-CD8 is useful for Flow Cytometry (Cell Surface). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.25 µg)
Do not freeze!

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: RPA-T8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR