Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT3]

Supplier: Rockland Immunochemical
200-326-M62
RL200-326-M62EA 355.22 USD
RL200-326-M62
Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT3]
Antibodies
Anti-CD3 is useful for Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/1x10^6 cells
Do not freeze!

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: OKT3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR