Anti-ITGAM Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HIB19]

Supplier: Rockland Immunochemical
200-308-N20
RL200-308-N20EA 470.15 USD
RL200-308-N20
Anti-ITGAM Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HIB19]
Antibodies
Anti-CD11b is useful for Flow Cytometry, Immunofluorescence, and Immunoprecipitation. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5µl/test/1x10e5 to 1x10e8 cells;
Do not freeze! Light sensitive!

Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: HIB19
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR