Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 14E5.A2.B2.H9]

Supplier: Rockland Immunochemical
200-306-I51
RL200-306-I51EA 547.09 USD
RL200-306-I51
Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 14E5.A2.B2.H9]
Antibodies
Anti-AKT3 BIOTIN Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT3 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9
Conjugation: Biotin
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR