Anti-6xHis Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 39A1413]

Supplier: Rockland Immunochemical
200-303-382
RL200-303-382EA 531.79 USD
RL200-303-382
Anti-6xHis Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 39A1413]
Antibodies
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 39A1413
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR