Anti-CD40L Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 24-31]

Supplier: Rockland Immunochemical
200-302-N76
RL200-302-N76EA 575.67 USD
RL200-302-N76
Anti-CD40L Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 24-31]
Antibodies
Anti-CD154 is useful for Flow Cytometry and Immunofluorescence. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells
Do not freeze!

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 24-31
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR