Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 6C12.A2.H4.A2.F9]

Supplier: Rockland Immunochemical
200-302-N62
RL200-302-N62EA 251.79 USD
RL200-302-N62
Anti-CD8A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 6C12.A2.H4.A2.F9]
Antibodies
Anti-CD8a is useful for immunochemistry and Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 6C12.A2.H4.A2.F9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR