Anti-UBB Mouse Monoclonal Antibody [clone: 5B9-B3]

Supplier: Rockland Immunochemical
200-301-G61 200-301-G62
RL200-301-G61EA 588.97 USD
RL200-301-G61 RL200-301-G62
Anti-UBB Mouse Monoclonal Antibody [clone: 5B9-B3]
Antibodies
Anti-Ubiquitin Antibody is recommended for use in WB and ELISA. Expect a band approximately ~10kDa corresponding to free ubiquitin. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000

Type: Primary
Antigen: UBB
Clonality: Monoclonal
Clone: 5B9-B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR