Anti-TRPV3 Mouse Monoclonal Antibody [clone: S15-4]

Supplier: Rockland Immunochemical
200-301-G58 200-301-G59
RL200-301-G58EA 588.97 USD
RL200-301-G58 RL200-301-G59
Anti-TRPV3 Mouse Monoclonal Antibody [clone: S15-4]
Antibodies
Anti-TrpV3 Antibody is suitable for use in WB, Immunofluorescence microscopy and IP. Expect a band approximately ~70kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: TRPV3
Clonality: Monoclonal
Clone: S15-4
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR