Anti-TRPC7 Mouse Monoclonal Antibody [clone: S64A-36]

Supplier: Rockland Immunochemical
200-301-G56
RL200-301-G56EA 588.97 USD
RL200-301-G56
Anti-TRPC7 Mouse Monoclonal Antibody [clone: S64A-36]
Antibodies
Anti-TrpC7 Antibody is suitable for use in WB, Immunochemistryand IP. Expect a band approximately ~100kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: TRPC7
Clonality: Monoclonal
Clone: S64A-36
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR