Anti-SHANK3 Mouse Monoclonal Antibody [clone: S69-46]

Supplier: Rockland Immunochemical
200-301-G45
RL200-301-G45EA 588.97 USD
RL200-301-G45
Anti-SHANK3 Mouse Monoclonal Antibody [clone: S69-46]
Antibodies
Anti-SHANK3 Antibody is suitable for use in WB, Immunochemistryand IP. Expect a band approximately ~190kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: SHANK3
Clonality: Monoclonal
Clone: S69-46
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR