Anti-MAPK14 Mouse Monoclonal Antibody [clone: 9F12]

Supplier: Rockland Immunochemical
200-301-G27
RL200-301-G27EA 588.97 USD
RL200-301-G27
Anti-MAPK14 Mouse Monoclonal Antibody [clone: 9F12]
Antibodies
Anti-p38 MAP kinase Antibody is recommended for use in WB, IP, and ELISA. Expect a band approximately ~38kDa Protein corresponding to the molecular mass of p38a MAPK on SDS PAGE immunoblots.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000;

Type: Primary
Antigen: MAPK14
Clonality: Monoclonal
Clone: 9F12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR