Anti-KCNA2 Mouse Monoclonal Antibody [clone: S14-16]

Supplier: Rockland Immunochemical
200-301-F92
RL200-301-F92EA 588.97 USD
RL200-301-F92
Anti-KCNA2 Mouse Monoclonal Antibody [clone: S14-16]
Antibodies
Anti-Kv1.2 K+ channel Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~80kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 1.0-10µg/ml; Western Blot: 1µg/ml; Immunochemistry: 0.1-1.0µg/ml

Type: Primary
Antigen: KCNA2
Clonality: Monoclonal
Clone: S14-16
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Zebrafish, Xenopus, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR